Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D630029K05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027846.1 | 2 | 240 | |||
NR_027847.1 | 2 | 350 | ||||
GenBank mRNA | AK085456.1 | - | 2 | 149 | ||
AW475548.1 | - | 2 | 303 | |||
BB843438.1 | - | 2 | 351 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CA481170.1 | - | 185 | Chr. - - [-] on Build |