Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snora3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028079.1 | 1 | 57 | |||
GenBank mRNA | AF357390.1 | - | 1 | 57 |
Gene Symbol: Rpl27a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK138110.1 | - | 2 | 632 |