Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Six3os1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_015385.2 | 2 | 556 | |||
NR_015386.2 | 2 | 465 | ||||
NR_015387.2 | 3 | 818 | ||||
NR_038082.1 | 2 | 465 | ||||
NR_038083.1 | 2 | 465 | ||||
NR_038084.1 | 2 | 465 | ||||
NR_038085.1 | 2 | 465 | ||||
NR_038086.1 | 2 | 465 | ||||
GenBank mRNA | AK044330.1 | - | LIT3371 RNCR1 |
2 | 2327 | |
AK053722.1 | - | 2 | 467 | |||
AY589790.1 | - | 2 | 344 | |||
AY589791.1 | - | 3 | 787 | |||
AY589792.1 | - | 1480 | ||||
AY590888.1 | - | 2 | 294 | |||
AY590889.1 | - | 2 | 384 | |||
BC065087.1 | - | 2 | 454 | |||
CA527570.1 | - | 2 | 409 | |||
CF737515.1 | - | 2 | 425 | |||
CF737925.1 | - | 2 | 441 | |||
CV783894.1 | - | 1 | 22 |