Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm13034 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030771.1 | 1 | 980 | |||
GenBank mRNA | AK141203.1 | - | 1 | 983 | ||
BC139113.1 | - | 1 | 972 | 8862135 |