Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbrg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027799.1 | 1 | 337 | |||
GenBank mRNA | AK042509.1 | - | 1 | 338 | ||
AK090121.1 | - | 1663 | ||||
BC095996.1 | - | 1 | 244 |