Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarca5-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002888.2 | 1 | 1137 | |||
GenBank mRNA | AK041999.1 | - | 1 | 1144 | ||
AK079035.1 | - | 1 | 214 |