Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxaas3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015611.2 | 3 | 716 | |||
GenBank mRNA | AK028207.1 | - | 3 | 717 | ||
AK146012.1 | - | 611 |