Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxaas3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015611.2 | 3 | 668 | |||
GenBank mRNA | AK012572.1 | - | 3 | 601 | ||
AK028207.1 | - | 3 | 669 | |||
AK146012.1 | - | 563 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY737288.1 | - | 3 | 563 | Chr. 6 - 52175233 - 52213529 [+] on Build GRCm38 |