Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxaas3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015611.2 | 1 | 48 | |||
GenBank mRNA | AK028207.1 | - | 1 | 49 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY711751.1 | - | 1 | 49 | Chr. 6 - 52201124 - 52213508 [+] on Build GRCm38 |