Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otx2os1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_029384.1 | 1 | 67 | |||
GenBank mRNA | AK042665.1 | - | LIT3409 Otx2OS |
1 | 67 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB639822.1 | - | 1 | 67 | Chr. 14 - 48669288 - 48793291 [+] on Build GRCm38 |