Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm14057 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024097.1 | 1 | 507 | |||
GenBank mRNA | AK013448.1 | - | 1 | 269 | ||
BC057040.1 | - | 2094 |