Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srrm4os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015595.2 | 4 | 637 | |||
GenBank mRNA | AK049160.1 | - | 4 | 638 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BF463123.1 | - | 1 | 94 | Chr. 5 - 116462676 - 116463161 [+] on Build GRCm38 |