Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700020I14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015473.1 | 3 | 1146 | |||
NR_027832.1 | 3 | 977 | ||||
GenBank mRNA | AK006166.1 | - | 3 | 1116 | ||
AK009929.1 | - | 3 | 953 | |||
AK011393.1 | - | 3 | 1110 | |||
AK017100.1 | - | 3 | 978 | |||
AK029148.1 | - | 1112 | ||||
AK030275.1 | - | 1109 | ||||
AK147270.1 | - | 1123 | ||||
BC026913.1 | - | 3 | 943 |