Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700020I14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015473.1 | 3 | 657 | |||
NR_027832.1 | 3 | 488 | ||||
GenBank mRNA | AK006166.1 | - | 3 | 627 | ||
AK009929.1 | - | 3 | 463 | |||
AK011393.1 | - | 3 | 621 | |||
AK011848.1 | - | 3 | 627 | |||
AK017100.1 | - | 3 | 489 | |||
AK017322.1 | - | 3 | 621 | |||
AK029148.1 | - | 623 | ||||
AK030275.1 | - | 621 | ||||
AK147270.1 | - | 634 | ||||
AK170765.1 | - | 3 | 627 | |||
BC026913.1 | - | 3 | 453 |