Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700020I14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015473.1 | 3 | 1267 | |||
NR_027832.1 | 3 | 1098 | ||||
GenBank mRNA | AK006166.1 | - | 3 | 1237 | ||
AK009929.1 | - | 3 | 1074 | |||
AK017100.1 | - | 3 | 1099 | |||
AK029148.1 | - | 1233 | ||||
AK030275.1 | - | 1230 | ||||
AK147270.1 | - | 1244 | ||||
BC026913.1 | - | 3 | 1064 |