Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rn28s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003279.1 | 34 |
Gene Symbol: Rn45s | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046233.2 | 8156 |