Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp326 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030777.1 | 3, 4 | 260 | |||
GenBank mRNA | BC037055.1 | - | 3, 4 | 265 | 4948371 | |
BU503985.1 | - | 2, 3 | 108 |