Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb44-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002879.2 | 2 | 190 | |||
GenBank mRNA | AY621334.1 | - | 2 | 190 | ||
DQ012049.1 | - | 2 | 190 |