Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4632415L05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027985.1 | 1 | 3623 | |||
GenBank mRNA | BC037267.1 | - | 1 | 3596 |