Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snora75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028478.1 | 1 | 84 | |||
GenBank mRNA | AF357416.1 | - | 1 | 84 |
Gene Symbol: Dpep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK089165.1 | - | 4 | 2969 |