Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4933400A11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003635.1 | 3 | 1088 | |||
GenBank mRNA | AK015979.1 | - | 2 | 955 | ||
AK016585.1 | - | 3 | 1089 | |||
BC107240.1 | - | 3 | 1006 | |||
BC107241.1 | - | 3 | 1010 |