Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbmx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029425.1 | 10 | 1413 | |||
GenBank mRNA | AJ237846.1 | - | 10 | 1342 | ||
AK087940.1 | - | 4 | 518 | |||
AK165006.1 | - | 10 | 1360 |