Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A530053G22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015565.2 | 2 | 1109 | |||
GenBank mRNA | AK080087.1 | - | 2 | 1110 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB220146.2 | - | 1 | 334 | Chr. 6 - 60279043 - 60279680 [-] on Build GRCm38 |