Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnm3os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002870.2 | 1 | 2747 | |||
GenBank mRNA | AK053654.1 | - | 1 | 1532 | ||
AK164873.1 | - | 1 | 575 | |||
AY751519.1 | - | 1 | 2707 |