Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Speer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001586.3 | 1 | 84 | |||
GenBank mRNA | AY151400.1 | - | 85 |
Gene Symbol: 4933402N22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK077088.1 | - | 153 |
Gene Symbol: Gm6455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003596.2 | 1 | 41 | |||
GenBank mRNA | BC048648.1 | - | 1 | 41 |