Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chpt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027477.1 | 8, 9 | 1380 | |||
GenBank mRNA | AK007964.1 | - | 8, 9 | 1300 |