Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vsig8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027644.1 | 1 | 29 | |||
GenBank mRNA | BC064106.1 | - | 1 | 29 | 1528324 |