Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm2518 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015538.1 | 1 | 1177 | |||
GenBank mRNA | AK041085.1 | - | 2 | 1073 | ||
AK077736.1 | - | 1 | 1174 | |||
AK082962.1 | - | 1 | 1176 | |||
AK085400.1 | - | 1 | 1149 |