Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prmt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024139.2 | 2 | 2684 | |||
GenBank mRNA | AK087551.1 | - | 2 | 2232 | ||
AK087608.1 | - | 2 | 2685 | |||
AK157132.1 | - | 3 | 2638 | |||
BC066221.1 | - | 2 | 2661 | 30436952 |