Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015606.3 | 2 | 1134 | |||
GenBank mRNA | AK035234.1 | - | 1 | 1643 | ||
AK035285.1 | - | 2 | 1265 | |||
AK051953.1 | - | 2 | 1012 | |||
AK161046.1 | - | 2 | 1136 |