Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oas1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003507.1 | 5 | 982 | |||
GenBank mRNA | AB067529.1 | - | 5 | 979 | ||
AF418004.1 | - | 5 | 979 | |||
AF466822.1 | - | 5 | 936 | |||
AK080152.1 | - | 5 | 972 |