Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps15a-ps6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BM207059.2 | - | 1 | 332 |
Gene Symbol: Rps15a-ps4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BM195459.2 | - | 1 | 332 |