Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003508.1 | 14 | 2232 | |||
GenBank mRNA | AK143831.1 | - | 14 | 2233 | ||
BC007127.1 | - | 14 | 2214 | 3591798 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE629286.1 | - | 2 | 463 | Chr. 16 - 97558679 - 97560748 [+] on Build GRCm38 |