Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2010016I18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033207.1 | 2 | 463 | |||
GenBank mRNA | AK040034.1 | - | 1 | 374 | ||
AK080971.1 | - | 2 | 464 | |||
AK140363.1 | - | 2 | 464 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB645704.1 | - | 2 | 464 | Chr. 3 - 106482910 - 106485913 [-] on Build GRCm38 |