Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldoart2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006425.1 | - | 1 | 564 | ||
AK016920.1 | - | 1 | 544 | |||
EF662062.1 | - | 1 | 491 |