Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028499.1 | 1 | 71 | |||
GenBank mRNA | CA491597.1 | - | 1 | 105 | ||
CB600958.1 | - | 1 | 74 |