Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700012D14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015573.2 | 3 | 480 | |||
GenBank mRNA | AK005898.1 | - | 3 | 482 |