Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 3110039I08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK034202.1 | - | 1 | 3019 | ||
AK034222.1 | - | 2 | 1603 | |||
AK141402.1 | - | 1956 |