Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024513.1 | 2 | 399 | |||
GenBank mRNA | AA414041.1 | - | 2 | 211 | ||
CO797249.1 | - | 2 | 428 |