Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 9930014A18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030696.1 | 1 | 1390 | |||
GenBank mRNA | AK036813.1 | - | 1 | 15 | ||
AK046554.1 | - | 1 | 1391 | |||
AK050709.1 | - | 1 | 1404 |