Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 9930014A18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030696.1 | 3 | 4850 | |||
GenBank mRNA | AK035741.1 | - | 1 | 2048 | ||
AK036813.1 | - | 2 | 3446 | |||
BC099672.1 | - | 1 | 1305 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_177166.1 | - | 2048 | Chr. - - [-] on Build |