Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930415L06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK132812.1 | - | 2 | 746 | ||
BC150985.1 | - | 1 | 697 | 9087898 | ||
BC150987.1 | - | 1 | 697 | 9087900 |