Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm8615 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028061.1 | 1 | 755 | |||
GenBank mRNA | CB237974.1 | - | 1 | 151 |
Gene Symbol: Gnpda1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF088903.1 | - | 7 | 930 | ||
AF160355.1 | - | 7 | 918 |