Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027491.1 | 1 | 188 | |||
GenBank mRNA | AA199406.1 | - | 1 | 188 | ||
AF359558.1 | - | 1 | 127 | |||
BX529138.1 | - | 1 | 197 |