Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_015351.1 | 626 | ||||
GenBank mRNA | U69888.1 | - | LIT1675 IPW |
626 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | U79154.1 | - | 1 | 15 | Chr. - - [-] on Build |