Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm1943 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002928.2 | 1 | 462 | |||
GenBank mRNA | AK170003.1 | - | 1 | 470 | ||
BC024407.1 | - | 1 | 463 |
Gene Symbol: Wdr70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014750.1 | - | 4 | 481 | ||
BC037184.1 | - | 4 | 485 | |||
BC051482.1 | - | 4 | 451 | |||
BC061236.1 | - | 4 | 484 |