Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700123L14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003643.1 | 1 | 1150 | |||
GenBank mRNA | AK131738.1 | - | 1 | 1151 | ||
BC026810.1 | - | 1 | 1043 | |||
BC141191.1 | - | 2 | 1115 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU945678.1 | - | 1 | 105 | Chr. 6 - 96164504 - 96165158 [-] on Build GRCm38 |