Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033241.1 | 16 | 2833 | |||
GenBank mRNA | AK147534.1 | - | 14 | 3749 | ||
BC003925.1 | - | 6 | 1162 | |||
BC053923.1 | - | 16 | 2724 |