Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700001G17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033199.1 | 1 | 857 | |||
GenBank mRNA | AK005592.1 | - | 1 | 857 | ||
AK008236.1 | - | 1 | 840 | |||
AK008325.1 | - | 1 | 840 | |||
BC132564.1 | - | 1 | 660 | |||
BC132566.1 | - | 1 | 660 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV204454.1 | - | 1 | 272 | Chr. 1 - 33670409 - 33670706 [+] on Build GRCm38 |