Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rian | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_028261.1 | 11 | 1427 | |||
GenBank mRNA | AB063319.1 | - | LIT1623 Rian |
1 | 186 | |
AF498294.1 | - | 1 | 204 | |||
AK017440.1 | - | 1 | 157 |